The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009761.3 | NP_033891.1 | 6 - 6 | 3045 | 80 |
GenBank mRNA | AK004667.1 | - | 6 - 6 | 2991 | 80 |
AK155040.1 | - | 6 - 6 | 3015 | 80 | |
AK168482.1 | - | 6 - 6 | 3048 | 80 | |
AK169391.1 | - | 6 - 6 | 3048 | 80 | |
AY057069.1 | - | 7 - 7 | 3064 | 80 |