Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lst1
Entrez Gene ID
16988
Gene Name
leukocyte specific transcript 1
Gene Aliases
B144
Chromosome Location
Chr.17: 35185095 - 35188440 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF000428.1 - 2 - 3 173 81

View Full Product Details