The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027568.2 | NP_081844.1 | 1 - 1 | 851 | 127 |
GenBank mRNA | AK014635.1 | - | 1 - 1 | 852 | 127 |