The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147073.1 | NP_667284.1 | 1 - 1 | 50 | 80 |
GenBank mRNA | BC104110.1 | - | 1 - 1 | 50 | 80 |
BC104111.1 | - | 1 - 1 | 50 | 80 |