Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim61
Entrez Gene ID
260296
Gene Name
tripartite motif-containing 61
Gene Aliases
2czf61, AU018704, C85201, E330039K03Rik, Rnf35
Chromosome Location
Chr.8: 65012976 - 65018688 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153110.4 NP_694750.3 1 - 2 235 93
GenBank mRNA BC064738.1 - 1 - 2 113 93

View Full Product Details