Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Skint11
Entrez Gene ID
230623
Gene Name
selection and upkeep of intraepithelial T cells 11
Gene Aliases
A630098G03Rik, Gm569
Chromosome Location
Chr.4: 114163384 - 114245028 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177669.4 NP_808337.2 3 - 4 99
GenBank mRNA AK042513.1 - 3 - 4 99

View Full Product Details