The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183312.3 | NP_899135.3 | 3 - 4 | 1114 | 121 |
GenBank mRNA | AJ579702.1 | - | 3 - 4 | 1065 | 121 |