Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ostn
Entrez Gene ID
239790
Gene Name
osteocrin
Gene Aliases
Ostc
Chromosome Location
Chr.16: 27306983 - 27351209 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198112.2 NP_932780.1 4 - 5 463 126
GenBank mRNA AY395730.1 - 4 - 5 463 126
AY573932.1 - 4 - 5 615 126

View Full Product Details