The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_203320.3 | NP_976065.1 | 3 - 4 | 361 | 99 |
GenBank mRNA | AK144158.1 | - | 3 - 4 | 363 | 99 |
AK150314.1 | - | 3 - 4 | 360 | 99 | |
AK154334.1 | - | 3 - 4 | 360 | 99 | |
AY311403.1 | - | 3 - 4 | 359 | 99 |