Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tas2r114
Entrez Gene ID
387346
Gene Name
taste receptor, type 2, member 114
Gene Aliases
T2R14, Tas2r14, mGR14, mt2r46
Chromosome Location
Chr.6: 131689134 - 131690063 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207019.1 NP_996902.1 1 - 1 767 151

View Full Product Details