The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207262.2 | NP_997145.2 | 1 - 2 | 73 | 85 |
GenBank mRNA | AY370634.1 | - | 1 - 2 | 81 | 85 |
CK616217.1 | - | 1 - 2 | 73 | 85 | |
GU269234.1 | - | 1 - 2 | 60 | 85 |