Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk1b26
Entrez Gene ID
16618
Gene Name
kallikrein 1-related petidase b26
Gene Aliases
Egfbp2, Klk26, PRECE-2, mGK-26, mGK26
Chromosome Location
Chr.7: 44007953 - 44016969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010644.2 NP_034774.1 1 - 2 86 115
GenBank mRNA K01831.1 - 1 - 2 86 115

View Full Product Details