Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Psme2
Entrez Gene ID
19188
Gene Name
proteasome (prosome, macropain) activator subunit 2 (PA28 beta)
Gene Aliases
AA589371, AI788882, PA28b
Chromosome Location
Chr.14: 55587440 - 55591146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011190.3 NP_035320.1 1 - 2 180 148
GenBank mRNA BC057859.1 - 1 - 2 177 148
BU563130.1 - 1 - 2 249 148
BY134400.1 - 1 - 2 146 148

View Full Product Details