The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282976.1 | NP_001269905.1 | 2 - 3 | 516 | 269 |
NM_013608.3 | NP_038636.2 | 2 - 3 | 405 | 158 | |
GenBank mRNA | AK146197.1 | - | 2 - 3 | 143 | 158 |
AK168023.1 | - | 2 - 3 | 118 | 158 | |
AK169432.1 | - | 2 - 3 | 177 | 158 | |
AL355707.1 | - | 2 - 3 | 138 | 158 | |
BU962928.1 | - | 2 - 3 | 379 | 158 | |
CN840834.1 | - | - | 408 | 158 | |
CX236470.1 | - | 2 - 3 | 241 | 269 |