Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hspa8
Entrez Gene ID
15481
Gene Name
heat shock protein 8
Gene Aliases
2410008N15Rik, Hsc70, Hsc71, Hsc73, Hsp73, Hspa10
Chromosome Location
Chr.9: 40801273 - 40805199 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC006722.1 - 1 - 2 96 120

View Full Product Details