Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdcd5
Entrez Gene ID
56330
Gene Name
programmed cell death 5
Gene Aliases
2200003D22Rik, AA408513, Tfar19
Chromosome Location
Chr.7: 35641985 - 35647482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019746.4 NP_062720.1 1 - 1 46 128
GenBank mRNA AK008632.1 - 1 - 1 60 128
BC092092.1 - 1 - 1 46 128

View Full Product Details