Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Csnka2ip
Entrez Gene ID
224291
Gene Name
casein kinase 2, alpha prime interacting protein
Gene Aliases
Ckt2
Chromosome Location
Chr.16: 64477810 - 64602366 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173861.2 NP_776286.1 1 - 1 1253 92
GenBank mRNA AF463503.1 - 1 - 1 1296 92
BC137818.1 - 1 - 1 1175 92

View Full Product Details