The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029529.3 | NP_083805.1 | 1 - 2 | 877 | 84 |
GenBank mRNA | AK042761.1 | - | 1 - 2 | 629 | 84 |
AK044973.1 | - | 1 - 2 | 614 | 84 | |
AK047610.1 | - | 1 - 2 | 878 | 84 | |
BC139194.1 | - | 1 - 2 | 707 | 84 |