Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Eed
- Entrez Gene ID
-
13626
- Gene Name
- embryonic ectoderm development
- Gene Aliases
-
ENSMUSG00000039373, l(7)5Rn, l7Rn5, lusk
- Chromosome Location
-
Chr.7: 89954654 - 89981419 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016228.1 |
- |
1 - 1 |
850 |
73 |
AK053947.1 |
- |
- |
844 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
lethal, Chr 7, Rinchik 6 |
l7Rn6 |
0610007P06Rik, 1110002N09Rik, Hikeshi, l(7)6Rn |
-39.5kb
|
Anti-sense |
Chr. - 89918685 - 89941221 on Build GRCm38 |
Downstream |
RIKEN cDNA E230029C05 gene |
E230029C05Rik |
AI452102, B230337J15 |
47.3kb
|
Sense |
Chr. - 90029159 - 90049071 on Build GRCm38 |
View Full Product Details