The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011496.1 | NP_035626.1 | 7 - 8 | 1190 | 68 |
GenBank mRNA | AK075951.1 | - | 7 - 8 | 1156 | 68 |
AK132006.1 | - | 7 - 8 | 1184 | 68 | |
BC003261.1 | - | 7 - 8 | 1182 | 68 | |
D21099.1 | - | 7 - 8 | 1055 | 68 | |
U69107.1 | - | - | 1133 | 68 |