Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4bp1
Entrez Gene ID
80750
Gene Name
NEDD4 binding protein 1
Gene Aliases
AI481586, C81621
Chromosome Location
Chr.8: 86841136 - 86885258 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK053991.1 - - 94 79

View Full Product Details