The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029300.1 | NP_083576.1 | 1 - 2 | 88 | 66 |
GenBank mRNA | AK005734.1 | - | 1 - 2 | 89 | 66 |
BC125546.1 | - | 1 - 2 | 47 | 66 |