Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc8e
Entrez Gene ID
72267
Gene Name
leucine rich repeat containing 8 family, member E
Gene Aliases
1810049O03Rik, C87354
Chromosome Location
Chr.8: 4224280 - 4237470 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028175.2 NP_082451.2 3 - 3 3823 101
GenBank mRNA BC031863.1 - 1 - 1 101

View Full Product Details