Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ccdc85b
Entrez Gene ID
240514
Gene Name
coiled-coil domain containing 85B
Gene Aliases
AI842788, BB115872
Chromosome Location
Chr.19: 5453163 - 5457563 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243307.1 NP_001230236.1 1 - 2 225 78
GenBank mRNA BC016659.1 - 1 - 2 160 78
BC094592.1 - 1 - 2 166 78

View Full Product Details