The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025264.1 | NP_001020435.1 | 5 - 6 | 500 | 109 |
GenBank mRNA | BE634665.1 | - | 5 - 6 | 482 | 109 |