Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nmnat1
- Entrez Gene ID
-
66454
- Gene Name
- nicotinamide nucleotide adenylyltransferase 1
- Gene Aliases
-
2610529L11Rik, 5730441G13Rik, D4Cole1e, nmnat
- Chromosome Location
-
Chr.4: 149468597 - 149485142 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK012194.1 |
- |
1 - 2 |
265 |
111 |
View Full Product Details