Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sdr39u1
Entrez Gene ID
654795
Gene Name
short chain dehydrogenase/reductase family 39U, member 1
Gene Aliases
2310014G06Rik
Chromosome Location
Chr.14: 55897285 - 55900281 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC058857.1 - 1 - 2 402 122

View Full Product Details