Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hjurp
- Entrez Gene ID
-
381280
- Gene Name
- Holliday junction recognition protein
- Gene Aliases
-
6430706D22Rik, A730008H23Rik, C330011F01Rik, Gm975, Mfleg1
- Chromosome Location
-
Chr.1: 88258856 - 88277633 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC004768.1 |
- |
1 - 2 |
207 |
168 |
View Full Product Details