Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Cyp2d13
- Entrez Gene ID
-
68444
- Gene Name
- cytochrome P450, family 2, subfamily d, polypeptide 13
- Gene Aliases
-
1300007K12Rik
- Chromosome Location
-
Chr.15: 82636750 - 82642045 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003552.1 |
|
3 - 4 |
590 |
142 |
GenBank mRNA |
AK004933.1 |
- |
- |
420 |
142 |
BC018344.1 |
- |
3 - 4 |
570 |
142 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BF384010.1 |
- |
3 - 4 |
569 |
|
Chr. 15 - 82640111 - 82642025 [-] on Build GRCm38 |
141 |
GenBankmRNA |
BI144949.1 |
- |
3 - 4 |
546 |
|
Chr. 15 - 82639353 - 82642001 [-] on Build GRCm38 |
142 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cytochrome P450, family 2, subfamily d, polypeptide 34 |
Cyp2d34 |
|
-19.2kb
|
Sense |
Chr. - 82615965 - 82620907 on Build GRCm38 |
Downstream |
cytochrome P450, family 2, subfamily d, polypeptide 37, pseudogene |
Cyp2d37-ps |
EG627860, Gm6800 |
48.3kb
|
Sense |
Chr. - 82688750 - 82690058 on Build GRCm38 |
View Full Product Details