The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003552.1 | 5 - 6 | 927 | 145 | |
GenBank mRNA | AK004933.1 | - | - | 757 | 145 |
BC018344.1 | - | 5 - 6 | 907 | 145 |