The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007763.3 | NP_031789.1 | 4 - 5 | 342 | 123 |
GenBank mRNA | AI323004.1 | - | 4 - 5 | 290 | 123 |
AK003075.1 | - | 4 - 5 | 304 | 123 | |
AK008269.1 | - | 4 - 5 | 304 | 123 | |
AK012068.2 | - | 4 - 5 | 299 | 123 | |
AK168305.1 | - | 4 - 5 | 303 | 123 | |
BC031922.1 | - | 4 - 5 | 287 | 123 | |
BC058606.1 | - | 3 - 4 | 1386 | 123 | |
BC064074.1 | - | 4 - 5 | 291 | 123 | |
BX511593.1 | - | 4 - 5 | 288 | 123 |