Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r67
Entrez Gene ID
620672
Gene Name
vomeronasal 2, receptor 67
Gene Aliases
EG620672
Chromosome Location
Chr.7: 85136240 - 85155902 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102579.1 NP_001096049.1 5 - 5 1607 95

View Full Product Details