The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278671.1 | NP_001265600.1 | 2 - 2 | 1166 | 144 |
GenBank mRNA | AK014702.1 | - | 2 - 2 | 1163 | 144 |
AK016973.1 | - | 2 - 2 | 1165 | 144 | |
BC139158.1 | - | 1 - 1 | 1111 | 144 | |
BC139160.1 | - | 1 - 1 | 1111 | 144 |