Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C3ar1
Entrez Gene ID
12267
Gene Name
complement component 3a receptor 1
Gene Aliases
AZ3B, C3AR, HNFAG09
Chromosome Location
Chr.6: 122847140 - 122856157 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009779.2 NP_033909.1 2 - 2 3290 143
GenBank mRNA AK080984.1 - 1 - 1 650 143
AK081010.1 - - 1617 143
U77460.1 - 2 - 2 3138 143

View Full Product Details