Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Defb45
Entrez Gene ID
433490
Gene Name
defensin beta 45
Gene Aliases
Defb27, OTTMUSG00000015859
Chromosome Location
Chr.2: 152593191 - 152596485 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037752.2 NP_001032841.1 2 - 2 271 124
GenBank mRNA AK005919.1 - 2 - 2 272 124
AY591384.1 - 2 - 2 216 124
BC132226.1 - 2 - 2 211 124
BC132228.1 - 2 - 2 211 124
DQ012038.1 - 2 - 2 268 124

View Full Product Details