Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubtfl1
Entrez Gene ID
546118
Gene Name
upstream binding transcription factor, RNA polymerase I-like 1
Gene Aliases
B020006M18Rik, Hmgpi
Chromosome Location
Chr.9: 18404418 - 18411502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033793.3 NP_001028965.1 3 - 3 1696 80
GenBank mRNA AK139937.1 - 3 - 3 1697 80
AK163257.1 - 2 - 2 1813 80
BC139141.1 - 2 - 2 1511 80
BC145822.1 - 2 - 2 1511 80

View Full Product Details