The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027629.3 | NP_081905.1 | 14 - 14 | 5284 | 151 |
GenBank mRNA | AK043357.1 | - | 1 - 1 | 1177 | 151 |
AK045551.1 | - | 1 - 1 | 2491 | 151 | |
AK164085.1 | - | 1 - 1 | 227 | 151 |