Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smad2
Entrez Gene ID
17126
Gene Name
SMAD family member 2
Gene Aliases
7120426M23Rik, Madh2, Madr2, Smad-2, mMad2
Chromosome Location
Chr.18: 76241251 - 76311748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252481.1 NP_001239410.1 11 - 11 8760 79
NM_010754.5 NP_034884.2 11 - 11 8649 79
GenBank mRNA AK078612.1 - 1 - 1 1551 79
AV232667.2 - 1 - 1 565 79
BM248478.2 - 1 - 1 79

View Full Product Details