Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Plekhf1
Entrez Gene ID
72287
Gene Name
pleckstrin homology domain containing, family F (with FYVE domain) member 1
Gene Aliases
1810013P09Rik, APPD, LAPF, PHAFIN1, ZFYVE15
Chromosome Location
Chr.7: 38220654 - 38227994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024413.2 NP_077724.2 2 - 2 577 91
GenBank mRNA AK143976.1 - - 566 91
AK157990.1 - 2 - 2 566 91
AK170283.1 - 2 - 2 576 91
AK171398.1 - 2 - 2 566 91
BC002120.1 - 1 - 1 542 91

View Full Product Details