Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vmn2r123
Entrez Gene ID
22300
Gene Name
vomeronasal 2, receptor 123
Gene Aliases
V2r1
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009485.1 NP_033511.1 2 - 3 542 77
GenBank mRNA AF011411.1 - 2 - 3 542 77

Gene Symbol
Vmn2r-ps159
Entrez Gene ID
22314
Gene Name
vomeronasal 2, receptor, pseudogene 159
Gene Aliases
V2r8, V2r9, Vmn2r-ps160
Chromosome Location
Chr.4: 156331103 - 156339499 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_028141.1 2 - 3 565 77
GenBank mRNA AF011419.1 - 2 - 3 565 77

View Full Product Details