The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026242.3 | NP_080518.1 | 1 - 2 | 559 | 76 |
GenBank mRNA | AB041651.1 | - | 1 - 2 | 541 | 76 |
AK005313.1 | - | 1 - 2 | 547 | 76 | |
AK009827.1 | - | 1 - 2 | 514 | 76 | |
AK017832.1 | - | 1 - 2 | 541 | 76 | |
AK018675.1 | - | 1 - 2 | 510 | 76 | |
AK159464.1 | - | 1 - 2 | 547 | 76 | |
AK167112.1 | - | 1 - 2 | 541 | 76 | |
AK170909.1 | - | 1 - 2 | 536 | 76 | |
AK170984.1 | - | 1 - 2 | 542 | 76 | |
BC010209.1 | - | 1 - 2 | 528 | 76 | |
BC085488.1 | - | 1 - 2 | 404 | 76 |