Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krt77
Entrez Gene ID
406220
Gene Name
keratin 77
Gene Aliases
4732484G22Rik, K1emb, Krt239, mK1b
Chromosome Location
Chr.15: 101858731 - 101869706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003667.1 NP_001003667.1 5 - 6 1092 139
GenBank mRNA AK132513.1 - 5 - 6 1180 139
AY379112.1 - 4 - 5 531 139
AY651826.1 - 4 - 5 531 139
BC107395.2 - 5 - 6 1092 139
BC107396.2 - 5 - 6 1092 139
BC141259.1 - 5 - 6 1115 139

View Full Product Details