Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rpl26
- Entrez Gene ID
-
19941
- Gene Name
- ribosomal protein L26
- Gene Aliases
-
SIG-20
- Chromosome Location
-
Chr.11: 68901566 - 68904537 on Build GRCm38
- Gene Symbol
- Gm15772
- Entrez Gene ID
-
100034726
- Gene Name
- ribosomal protein L26 pseudogene
- Gene Aliases
-
ENSMUSG00000062353, OTTMUSG00000026108
- Chromosome Location
-
Chr.5: 3236390 - 3236928 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003373.1 |
|
1 - 1 |
352 |
165 |
GenBank mRNA |
AK146116.1 |
- |
1 - 1 |
351 |
165 |
X80699.1 |
- |
1 - 1 |
358 |
165 |
View Full Product Details