The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010114.1 | NP_034244.1 | 4 - 5 | 642 | 159 |
GenBank mRNA | BC106155.1 | - | 4 - 5 | 646 | 159 |