The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010185.4 | NP_034315.1 | 3 - 4 | 288 | 137 |
GenBank mRNA | AK155600.1 | - | 3 - 4 | 245 | 137 |
BC034163.1 | - | - | 228 | 137 | |
BY173505.1 | - | 3 - 4 | 288 | 137 |