Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfx
Entrez Gene ID
22764
Gene Name
zinc finger protein X-linked
Gene Aliases
Zfx5, Zfx5,6, Zfx6
Chromosome Location
Chr.X: 94074631 - 94150643 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044386.1 NP_001037851.1 5 - 6 1217 144
NM_011768.2 NP_035898.2 4 - 5 1149 144
GenBank mRNA BC137859.1 - 3 - 4 817 144
CJ195389.1 - 1 - 2 67 144
M32308.1 - 5 - 6 925 144
M32309.1 - 4 - 5 978 144

View Full Product Details