Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqsec2
Entrez Gene ID
245666
Gene Name
IQ motif and Sec7 domain 2
Gene Aliases
Brag1, mKIAA0522
Chromosome Location
Chr.X: 152144183 - 152225237 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005475.2 NP_001005475.1 12 - 13 2718 146
NM_001114664.2 NP_001108136.1 12 - 13 3455 146
GenBank mRNA AB275281.1 - 12 - 13 3455 146
AK220345.1 - 6 - 7 753 146
AY451401.1 - 12 - 13 3478 146

View Full Product Details