Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Iqsec2
Entrez Gene ID
245666
Gene Name
IQ motif and Sec7 domain 2
Gene Aliases
Brag1, mKIAA0522
Chromosome Location
Chr.X: 152144183 - 152225237 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005475.2 NP_001005475.1 2 - 3 178 140

View Full Product Details