Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirpb1a
Entrez Gene ID
320832
Gene Name
signal-regulatory protein beta 1A
Gene Aliases
9930027N05Rik, SIRP-beta, Sirpb, Sirpb1
Chromosome Location
Chr.3: 15371661 - 15426460 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002898.1 NP_001002898.1 4 - 5 1087 104
GenBank mRNA AB109404.1 - - 1164 104
AB112022.1 - 4 - 5 1087 104
AB112024.1 - - 1124 104

Gene Symbol
LOC100038947
Entrez Gene ID
100038947
Gene Name
signal-regulatory protein beta 1-like
Gene Aliases
-
Chromosome Location
Chr.3: 15795147 - 15848530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173459.2 NP_001166930.1 4 - 5 1139 104
GenBank mRNA AK029805.1 - 1 - 2 210 104
BY551146.1 - 1 - 2 123 104
DQ055453.1 - 4 - 5 1088 104

Gene Symbol
Sirpb1b
Entrez Gene ID
668101
Gene Name
signal-regulatory protein beta 1B
Gene Aliases
-
Chromosome Location
Chr.3: 15495754 - 15575065 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173460.1 NP_001166931.1 4 - 5 1180 104
GenBank mRNA AK154879.1 - 4 - 5 1181 104

View Full Product Details