Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Vmn2r43
- Entrez Gene ID
-
381838
- Gene Name
- vomeronasal 2, receptor 43
- Gene Aliases
-
EC2-V2R, Gm1961
- Chromosome Location
-
Chr.7: 8244348 - 8260599 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198961.2 |
NP_945199.1 |
1 - 2 |
237 |
133 |
GenBank mRNA |
AJ543405.1 |
- |
1 - 2 |
239 |
133 |
- Gene Symbol
- Vmn2r35
- Entrez Gene ID
-
625353
- Gene Name
- vomeronasal 2, receptor 35
- Gene Aliases
-
EG625353
- Chromosome Location
-
Chr.7: 7786151 - 7819867 on Build GRCm38
- Gene Symbol
- Vmn2r39
- Entrez Gene ID
-
545909
- Gene Name
- vomeronasal 2, receptor 39
- Gene Aliases
-
EG545909
- Chromosome Location
-
Chr.7: 9014750 - 9030682 on Build GRCm38
- Gene Symbol
- Vmn2r50
- Entrez Gene ID
-
434117
- Gene Name
- vomeronasal 2, receptor 50
- Gene Aliases
-
EG434117
- Chromosome Location
-
Chr.7: 10037235 - 10053178 on Build GRCm38
View Full Product Details